Daidzein ≥98%

Supplier: Apollo Scientific
Warning

Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one

BIMB2005-1G BIMB2005-250MG
APOSBIMB2005-1GEA 87.3 EUR
APOSBIMB2005-1G APOSBIMB2005-250MG
Daidzein ≥98%
Daidzein

Inactive analogue of the tyrosine kinase inhibitor Genistein.

Formula: C₁₅H₁₀O₄
MW: 254,24 g/mol
Melting Pt: 328…333 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00016954
CAS Number: 486-66-8
EINECS: 207-635-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR