Naphthol AS-TR phosphate disodium salt

Supplier: Apollo Scientific
Warning

BIN0365-1G BIN0365-500MG BIN0365-1680G BIN0365-240G BIN0365-480G
APOSBIN0365-1GEA 55.3 EUR
APOSBIN0365-1G APOSBIN0365-500MG APOSBIN0365-1680G APOSBIN0365-240G APOSBIN0365-480G
Naphthol AS-TR phosphate disodium salt
Naphthol AS-TR phosphate disodium salt

Substrate for the histochemical demonstration of acid and alkaline phosphatase

Formula: C₁₈H₁₃ClNNa₂O₅P
Storage Temperature: Freezer
CAS Number: 4264-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR