Oligomycin B (from Streptomyces diastatochromogenes)

Supplier: Apollo Scientific
Warning

BIO1004-10MG BIO1004-1MG BIO1004-5MG
APOSBIO1004-10MGEA 727 EUR
APOSBIO1004-10MG APOSBIO1004-1MG APOSBIO1004-5MG
Oligomycin B (from Streptomyces diastatochromogenes)
Oligomycin B

Practically free of homologs.

Formula: C₄₅H₇₂O₁₂
Storage Temperature: Freezer
CAS Number: 11050-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR