Oligomycin C
Supplier: Apollo Scientific
|
Warning
|
BIO1007-5MG
BIO1007-1MG
APOSBIO1007-5MGEA
698
EUR
APOSBIO1007-5MG
APOSBIO1007-1MG
Oligomycin C
Oligomycin C
Formula:
C₄₅H₇₄O₁₀ Storage Temperature: Freezer |
CAS Number:
11052-72-5 EINECS: 234-276-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...