PIPES sesquisodium salt ≥96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid sesquisodium salt, Sesquisodium piperazine-1,4-diethanesulphonate, Sodium 1,1'-(piperazine-1,4-diyl)diethanesulphonate 1-(4-(1-sulphoethyl)piperazin-1-yl)ethanesulphonate(3:2), PIPES-Na1,5, Piperazine-N,N'-bis-2-ethanesulphonic acid sesquisodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) sesquisodium salt
BIP1706-100G
BIP1706-500G
APOSBIP1706-100GEA
62.6
EUR
APOSBIP1706-100G
APOSBIP1706-500G
PIPES sesquisodium salt ≥96%
PIPES sesquisodium salt
- Buffer
Formula:
C₁₆H₃₃N₄Na₃O₁₂S₄ MW: 670,69 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066393 CAS Number: 100037-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...