Paclobutrazol

Supplier: Apollo Scientific
Danger

BIP7675-100G BIP7675-25G BIP7675-5G
APOSBIP7675-100GEA 109 EUR
APOSBIP7675-100G APOSBIP7675-25G APOSBIP7675-5G
Paclobutrazol
Paclobutrazol
Formula: C₁₅H₂₀ClN₃O
Melting Pt: 165…166 °C
Storage Temperature: Ambient
MDL Number: MFCD01678673
CAS Number: 76738-62-0
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR