Amino-PEG8-acid

Supplier: Apollo Scientific
Warning

BIPG1036-100MG BIPG1036-50MG
APOSBIPG1036-100MGEA 46.4 EUR
APOSBIPG1036-100MG APOSBIPG1036-50MG
Amino-PEG8-acid
Amino-PEG8-acid
Formula: C₁₉H₃₉NO₁₀
MW: 441,51 g/mol
Storage Temperature: Ambient
CAS Number: 756526-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR