Amino-PEG4-alcohol

Supplier: Apollo Scientific
Warning

BIPG1046-10G BIPG1046-1G BIPG1046-5G
APOSBIPG1046-10GEA 683 EUR
APOSBIPG1046-10G APOSBIPG1046-1G APOSBIPG1046-5G
Amino-PEG4-alcohol
Amino-PEG4-alcohol
Formula: C₈H₁₉NO₄
Storage Temperature: Freezer
CAS Number: 86770-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR