Amino-PEG8-alcohol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Amino-dPEG®8-alcohol, NH2-PEG8-OH, 23-Amino-3,6,9,12,15,18,21-heptaoxatricosan-1-ol
BIPG1050-100MG
BIPG1050-250MG
BIPG1050-500MG
APOSBIPG1050-100MGEA
62.5
EUR
APOSBIPG1050-100MG
APOSBIPG1050-250MG
APOSBIPG1050-500MG
Amino-PEG8-alcohol
Amino-PEG8-alcohol
Formula:
C₁₆H₃₅NO₈ MW: 369.45 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11041128 CAS Number: 352439-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...