Azido-PEG24-acid
Supplier: Apollo Scientific
|
Warning
|
BIPG1113-100MG
BIPG1113-50MG
APOSBIPG1113-100MGEA
569
EUR
APOSBIPG1113-100MG
APOSBIPG1113-50MG
Azido-PEG24-acid
Azido-PEG24-acid
Formula:
C₅₁H₁₀₁N₃O₂₆ MW: 1172,35 g/mol Storage Temperature: Freezer |
CAS Number:
1167575-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...