Biotin-PEG2-Amine

Supplier: Apollo Scientific
Warning

Synonyms: N-(2-(2-(2-Aminoethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Biotin-DADOO, N-Biotinyl-3,6-dioxaoctane-1,8-diamine, N-[2-[2-(2-Aminoethoxy)ethoxy]ethyl]biotinamide

BIPG1207-100MG BIPG1207-50MG BIPG1207-250MG
APOSBIPG1207-100MGEA 217 EUR
APOSBIPG1207-100MG APOSBIPG1207-50MG APOSBIPG1207-250MG
Biotin-PEG2-Amine
Biotin-PEG2-Amine
Formula: C₁₆H₃₀N₄O₄S
Storage Temperature: Refrigerator
MDL Number: MFCD09264745
CAS Number: 138529-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR