(Fmoc-amino)-PEG12-C2-Carboxylic acid

Supplier: Apollo Scientific

Synonyms: Carboxy-PEG12-(Fmoc-amine), 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28,31,34,37,40-tridecaoxa-4-azatritetracontan-43-oic acid, Fmoc-NH-PEG12-CH2CH2COOH, FmocNH-PEG12-CH2CH2COOH

BIPG1405-100MG BIPG1405-50MG
APOSBIPG1405-100MGEA 401 EUR
APOSBIPG1405-100MG APOSBIPG1405-50MG
(Fmoc-amino)-PEG12-C2-Carboxylic acid
(Fmoc-amino)-PEG12-C2-Carboxylic acid
Formula: C₄₂H₆₅NO₁₆
MW: 839,97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064302
CAS Number: 1952360-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR