MAL-PEG8-acid

Supplier: Apollo Scientific

BIPG1475-100MG BIPG1475-250MG
APOSBIPG1475-100MGEA 69.6 EUR
APOSBIPG1475-100MG APOSBIPG1475-250MG
MAL-PEG8-acid
Mal-amido-PEG8-acid
Formula: C₂₆H₄₄N₂O₁₃
Storage Temperature: Ambient
MDL Number: MFCD13184956
CAS Number: 1334177-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR