Azido-PEG11-Amine

Supplier: Apollo Scientific

Synonyms: 35-Amino-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontanyl Azide, 35-Azido-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontan-1-amine, Amino-PEG11-Azide, 1-Amino-35-azido-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontane

BIPG1886-100MG BIPG1886-1G BIPG1886-500MG
APOSBIPG1886-100MGEA 647 EUR
APOSBIPG1886-100MG APOSBIPG1886-1G APOSBIPG1886-500MG
Azido-PEG11-Amine
Azido-PEG11-Amine
Formula: C₂₄H₅₀N₄O₁₁
MW: 570.68 g/mol
Storage Temperature: Ambient
CAS Number: 1800414-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR