N-Phthalyl-L-tryptophan ≥98%

Supplier: Apollo Scientific
Warning

Synonyms: (S)-2-(1,3-Dioxoisoindolin-2-yl)-3-(1H-indol-3-yl)propanoic acid, RG 108, RG108

BIR1004-10MG BIR1004-25MG
APOSBIR1004-10MGEA 21.9 EUR
APOSBIR1004-10MG APOSBIR1004-25MG
N-Phthalyl-L-tryptophan ≥98%
N-Phthalyl-L-tryptophan

DNA methyltransferase inhibitor. Reactivates tumour suppressor in gene expression in tumour cells.

Formula: C₁₉H₁₄N₂O₄
MW: 334,33 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08705332
CAS Number: 48208-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR