3-Hydroxy-6H-benzo[c]chromen-6-one 99%

Supplier: Apollo Scientific

Synonyms: Urolithin B

BISN0017-250MG BISN0017-1G
APOSBISN0017-250MGEA 58 EUR
APOSBISN0017-250MG APOSBISN0017-1G
3-Hydroxy-6H-benzo[c]chromen-6-one 99%
3-Hydroxy-6H-benzo[c]chromen-6-one

Urolithin B is a natural product with anti-proliferative and antioxidant activity, which can penetrate the blood-brain barrier and may have neuroprotective effects against Alzheimer’s Disease.

Formula: C₁₃H₈O₃
MW: 212,21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00034338
CAS Number: 1139-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR