Cryptotanshinone

Supplier: Apollo Scientific

Synonyms: (R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione, (R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethyl-phenanthro(1,2-b)furan-10,11-dione, Tanshinone C

BISN0122-10MG BISN0122-25MG BISN0122-50MG
APOSBISN0122-10MGEA 80 EUR
APOSBISN0122-10MG APOSBISN0122-25MG APOSBISN0122-50MG
Cryptotanshinone
Cryptotanshinone

Cryptotanshinone is a potent STAT3 inhibitor IC50 46 M and inhibits STAT3 Tyr705 phosphorylation in DU145 prostate cancer cells.

Formula: C₁₉H₂₀O₃
MW: 296,37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07636810
CAS Number: 35825-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR