Sivelestat sodium

Supplier: Apollo Scientific

Synonyms: ONO-5046, Sodium 2-(2-(4-(pivaloyloxy)phenylsulphonamido)benzamido)acetate tetrahydrate, N-{2-[({4-[2,2-Dimethylpropanoyl)oxy]phenyl}sulphonyl)amino]benzoyl}glycine sodium salt, Sivelestat Sodium Tetrahydrate

BISN0123-5MG BISN0123-10MG BISN0123-50MG
APOSBISN0123-5MGEA 78.3 EUR
APOSBISN0123-5MG APOSBISN0123-10MG APOSBISN0123-50MG
Sivelestat sodium
Sivelestat sodium

Sivelestat sodium is a competitive inhibitor of human neutrophil elastase (IC50 44 nM Ki200 nM) also inhibits leukocyte elastase obtained from rabbit, rat, hamster and mouse.

Formula: C₂₀H₂₁N₂NaO₇S·4H₂O
MW: 528,5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01937969
CAS Number: 201677-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR