DAPT

Supplier: Apollo Scientific

Synonyms: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester

BISN0140-5MG BISN0140-10MG BISN0140-25MG
APOSBISN0140-5MGEA 109 EUR
APOSBISN0140-5MG APOSBISN0140-10MG APOSBISN0140-25MG
DAPT
DAPT

DAPT is an inhibitor of -secretase and it can reduce the expression of A40 and A42 with IC50 values of 115 nM and 200 nM.

Formula: C₂₃H₂₆F₂N₂O₄
MW: 432,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974585
CAS Number: 208255-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR