Hydroxychloroquine sulphate

Supplier: Apollo Scientific

Synonyms: 2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol sulphate, N4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine sulphate, 7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline Sulphate

BISN0145-25G
APOSBISN0145-25GEA 78.3 EUR
APOSBISN0145-25G
Hydroxychloroquine sulphate
Hydroxychloroquine sulphate

Hydroxychloroquine sulfate is an antimalarial agent used for the treatment of systemic lupus erythematosus rheumatoid arthritis and other autoimmune inflammatory and dermatologic conditions Also acts as an inhibitor of autophagy and toll-like receptor TLR79.

Formula: C₁₈H₂₆ClN₃O
MW: 433,95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078203
CAS Number: 747-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR