β-Lapachone
Supplier: Apollo Scientific
BISN0149-5MG
BISN0149-10MG
BISN0149-50MG
APOSBISN0149-5MGEA
48.6
EUR
APOSBISN0149-5MG
APOSBISN0149-10MG
APOSBISN0149-50MG
β-Lapachone
beta-Lapachone
Lapachone is a naturally occurring quinone obtained from the bark of the lapacho tree (Tabebuia avellanedae) with cancer chemopreventive properties and a potent inhibitor of IDO1 (IC500.44 M).
Formula:
C₁₅H₁₄O₃ Melting Pt: 151…153 °C Storage Temperature: Freezer |
MDL Number:
MFCD01712233 CAS Number: 4707-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...