CH5424802 (Alectinib)
Supplier: Apollo Scientific
Synonyms:
Alectinib, 9-Ethyl-6,6-dimethyl-8-(4-morpholinopiperidin-1-yl)-11-oxo-6,11-dihydro-5H-benzo[b]carbazole-3-carbonitrile, 9-Ethyl-6,6-dimethyl-8-(4-morpholin-4-ylpiperidin-1-yl)-11-oxo-5H-benzo[b]carbazol-3-carbonitrile
BISN0151-2MG
BISN0151-5MG
BISN0151-10MG
APOSBISN0151-2MGEA
94.5
EUR
APOSBISN0151-2MG
APOSBISN0151-5MG
APOSBISN0151-10MG
CH5424802 (Alectinib)
CH5424802 (Alectinib)
Alectinib is a potent ALK inhibitor with IC50 of 1.9 nM while also inhibits F1174L, R1275Q, and L1196M mutants of ALK (IC50s 1.0, 3.5, and 1.6 nM, respectively).
Formula:
C₃₀H₃₄N₄O₂ MW: 482,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19440988 CAS Number: 1256580-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...