Ethyl-3-amino-4-(cyclohexylamino)benzoate

Supplier: Apollo Scientific

Synonyms: Ferrostatin-1, Ethyl 3-amino-4-(cyclohexylamino)benzoate

BISN0152-1MG
APOSBISN0152-1MGEA 0 EUR
APOSBISN0152-1MG
Ethyl-3-amino-4-(cyclohexylamino)benzoate
Ethyl-3-amino-4-(cyclohexylamino)benzoate

Ferrostatin-1 is a potent and selective inhibitor of ferroptosis with EC50 of 60 nM.

Formula: C₁₅H₂₂N₂O₂
MW: 262,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08072959
CAS Number: 347174-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR