DP-4978

Supplier: Apollo Scientific

Synonyms: 1-(3,3-dimethylbutyl)-3-(2-fluoro-4-methyl-5-(7-methyl-2-(methylamino)pyrido[2,3-d]pyrimidin-6-yl)phenyl)urea, LY3009120

BISN0175-5MG BISN0175-10MG BISN0175-25MG
APOSBISN0175-5MGEA 80 EUR
APOSBISN0175-5MG APOSBISN0175-10MG APOSBISN0175-25MG
DP-4978
DP-4978

MAPK/ERK Signalling Pathway

Formula: C₂₃H₂₉FN6O
MW: 424,52 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28411374
CAS Number: 1454682-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR