BI-78D3
Supplier: Apollo Scientific
Synonyms:
BI 78D3, 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-[(5-nitro-1,3-thiazol-2-yl)sulphanyl]-1H-1,2,4-triazol-5-one, BI78D3, 4-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-5-((5-nitrothiazol-2-yl)thio)-4H-1,2,4-triazol-3-ol, 4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-3-((5-nitrothiazol-2-yl)thio)-1H-1,2,4-triazol-5(4H)-one, BI-78D3
BISN0178-1MG
APOSBISN0178-1MGEA
36.4
EUR
APOSBISN0178-1MG
BI-78D3
BI-78D3
MAPK/ERK Signalling Pathway
Formula:
C₁₃H₉N₅O₅S₂ MW: 379,37 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01313575 CAS Number: 883065-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...