JNK-IN-8
Supplier: Apollo Scientific
BISN0191-2MG
BISN0191-5MG
BISN0191-10MG
APOSBISN0191-2MGEA
102
EUR
APOSBISN0191-2MG
APOSBISN0191-5MG
APOSBISN0191-10MG
JNK-IN-8
JNK-IN-8
MAPK/ERK Signalling Pathway
Formula:
C₂₉H₂₉N₇O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD22124890 CAS Number: 1410880-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...