LJI-308

Supplier: Apollo Scientific

Synonyms: 2,6-Difluoro-4-(4-(4-morpholinophenyl)pyridin-3-yl)phenol, 2,6-Difluoro-4-[4-[4-(4-morpholinyl)phenyl]-3-pyridinyl]-phenol

BISN0194-2MG BISN0194-5MG BISN0194-10MG
APOSBISN0194-2MGEA 174 EUR
APOSBISN0194-2MG APOSBISN0194-5MG APOSBISN0194-10MG
LJI-308
LJI-308

MAPK/ERK Signalling Pathway

Formula: C₂₁H₁₈F₂N₂O₂
MW: 368,38 g/mol
Storage Temperature: Freezer
MDL Number: MFCD29905461
CAS Number: 1627709-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR