Tyrphostin AG-126

Supplier: Apollo Scientific

Synonyms: 2-[(3-Hydroxy-4-nitrophenyl)methylidene]propanedinitrile, Tyrphostin AG 126, 2-(3-Hydroxy-4-nitrobenzylidene)malononitrile, AG-126, (3-Hydroxy-4-nitrobenzylidene)malononitrile

BISN0202-5MG
APOSBISN0202-5MGEA 58 EUR
APOSBISN0202-5MG
Tyrphostin AG-126
Tyrphostin AG-126

MAPK/ERK Signalling Pathway

Formula: C₁₀H₅N₃O₃
MW: 215,168 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236448
CAS Number: 118409-62-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR