PD 184,352

Supplier: Apollo Scientific

Synonyms: 2-(2-Chloro-4-iodoanilino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, 2-((2-Chloro-4-iodophenyl)amino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, PD 184352, CI-1040

BISN0210-5MG BISN0210-10MG BISN0210-50MG
APOSBISN0210-5MGEA 94.5 EUR
APOSBISN0210-5MG APOSBISN0210-10MG APOSBISN0210-50MG
PD 184,352
PD 184,352

MAPK/ERK Signalling Pathway

Formula: C₁₇H₁₄ClF₂IN₂O₂
MW: 478,7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683961
CAS Number: 212631-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR