Ralimetinib dimesylate

Supplier: Apollo Scientific

BISN0222-1MG BISN0222-5MG BISN0222-10MG
APOSBISN0222-1MGEA 80 EUR
APOSBISN0222-1MG APOSBISN0222-5MG APOSBISN0222-10MG
Ralimetinib dimesylate
LY2228820

MAPK/ERK Signalling Pathway

Formula: C₂₆H₃₇FN₆O₆S₂
Storage Temperature: Ambient
MDL Number: MFCD28101032
CAS Number: 862507-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR