AS601245
Supplier: Apollo Scientific
Synonyms:
2-(Benzo[d]thiazol-2-yl)-2-(2-((2-(pyridin-3-yl)ethyl)amino)pyrimidin-4-yl)acetonitrile
BISN0259-1MG
BISN0259-5MG
BISN0259-25MG
APOSBISN0259-1MGEA
72.7
EUR
APOSBISN0259-1MG
APOSBISN0259-5MG
APOSBISN0259-25MG
AS601245
AS601245
MAPK/ERK Signalling Pathway
Formula:
C₂₀H₁₆N₆S MW: 372,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07772196 CAS Number: 345987-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...