SD-06

Supplier: Apollo Scientific

BISN0285-1MG BISN0285-5MG BISN0285-10MG
APOSBISN0285-1MGEA 37.7 EUR
APOSBISN0285-1MG APOSBISN0285-5MG APOSBISN0285-10MG
SD-06
SD-06

MAPK/ERK Signalling Pathway

Formula: C₂₀H₂₀ClN₅O₂
Storage Temperature: Ambient
MDL Number: MFCD17019341
CAS Number: 271576-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR