D-(+)-Tyrosine ≥99%
Supplier: Apollo Scientific
|
Warning
|
BIT0722-100G
BIT0722-25G
APOSBIT0722-100GEA
73
EUR
APOSBIT0722-100G
APOSBIT0722-25G
D-(+)-Tyrosine ≥99%
D(+)-Tyrosine
Formula:
C₉H₁₁NO₃ Melting Pt: 310…314 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063073 CAS Number: 556-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...