Trichostatin A (from Streptomyces species)
Supplier: Apollo Scientific
|
Warning
|
BIT1005-10MG
BIT1005-5MG
APOSBIT1005-10MGEA
435
EUR
APOSBIT1005-10MG
APOSBIT1005-5MG
Trichostatin A (from Streptomyces species)
Trichostatin A
Formula:
C₁₇H₂₂N₂O₃ Storage Temperature: Freezer |
MDL Number:
MFCD03848392 CAS Number: 58880-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...