Trizma(R) maleate ≥98%

Supplier: Apollo Scientific
Warning

BIT3097-100G BIT3097-50G
APOSBIT3097-100GEA 87 EUR
APOSBIT3097-100G APOSBIT3097-50G
Trizma(R) maleate ≥98%
Trizma(R) maleate

  • Buffer

Formula: C₈H₁₅NO₇
Melting Pt: 104…106 °C
Storage Temperature: Ambient
CAS Number: 72200-76-1
EINECS: 276-455-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR