Virginiamycin S1

Supplier: Apollo Scientific

BIV1004-10MG BIV1004-5MG
APOSBIV1004-10MGEA 284 EUR
APOSBIV1004-10MG APOSBIV1004-5MG
Virginiamycin S1
Virginiamycin S1
Formula: C₄₃H₄₉N₇O₁₀
MW: 823.9 g/mol
Melting Pt: 165…176 °C
Storage Temperature: Freezer
CAS Number: 23152-29-6
EINECS: 245-462-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR