XTT sodium salt

Supplier: Apollo Scientific
Warning

Synonyms: 2,3-Bis(2-methoxy-4-nitro-5-sulphophenyl)-2H-tetrazolium-5-carboxanilide, XTT sodium, Sodium 5,5'-(5-(phenylcarbamoyl)-2H-tetrazole-3-ium-2,3-diyl)bis(4-methoxy-2-nitrobenzenesulphonate), sodium 2,3,-bis(2-methoxy-4-nitro-5-sulphophenyl)-5-[(phenylamino)-carbonyl]-2H-tetrazolium inner salt

BIX1003-100MG BIX1003-500MG
APOSBIX1003-100MGEA 314 EUR
APOSBIX1003-100MG APOSBIX1003-500MG
XTT sodium salt
XTT sodium salt

Useful in AIDS and cancer testing

Formula: C₂₂H₁₇N₇O₁₃S₂Na
MW: 673,52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083517
CAS Number: 111072-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR