Aluminium nitride 99,9%, powder ~325 mesh
Supplier: Apollo Scientific
Synonyms:
Aluminum nitride
IN1030-250G
IN1030-50G
IN1030-1KG
APOSIN1030-250GEA
248
EUR
APOSIN1030-250G
APOSIN1030-50G
APOSIN1030-1KG
Aluminium nitride 99,9%, powder ~325 mesh
Aluminium nitride
Formula:
AlN MW: 40,99 g/mol Melting Pt: >2200 °C Density: 3,26 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003429 CAS Number: 24304-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...