Barium wolframate 99,9%
Supplier: Apollo Scientific
Synonyms:
Barium(II) tungstate(VI)
IN1228-100G
IN1228-50G
APOSIN1228-100GEA
408
EUR
APOSIN1228-100G
APOSIN1228-50G
Barium wolframate 99,9%
Barium wolframate
Formula:
BaO₄W Density: 5,04 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS Number:
7787-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...