Calcium nitrate hydrate 99.99%

Supplier: Apollo Scientific
Danger

IN1390-25G IN1390-5G
APOSIN1390-25GEA 72.7 EUR
APOSIN1390-25G APOSIN1390-5G
Calcium nitrate hydrate 99.99%
Calcium nitrate hydrate
Formula: Ca(NO₃)₂·H₂O
MW: 182,1 g/mol
Boiling Pt: 132 °C
Melting Pt: 39,7 °C
Density: 1,82 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 35054-52-5
UN: 1454
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR