Dysprosium(III) sulphate octahydrate 99,9%
Supplier: Apollo Scientific
|
Warning
|
IN1669-100G
IN1669-25G
APOSIN1669-100GEA
415
EUR
APOSIN1669-100G
APOSIN1669-25G
Dysprosium(III) sulphate octahydrate 99,9%
Dysprosium (III) sulphate octahydrate
Formula:
Dy₂O₁₂S₃·8H₂O Storage Temperature: Ambient |
CAS Number:
10031-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...