Europium(III) chloride, anhydrous 99,9%
Supplier: Apollo Scientific
|
Warning
|
IN1765-25G
IN1765-5G
APOSIN1765-25GEA
1130
EUR
APOSIN1765-25G
APOSIN1765-5G
Europium(III) chloride, anhydrous 99,9%
Europium (III) chloride
Formula:
Cl₃Eu MW: 258,32 g/mol Melting Pt: 850 °C Density: 4,89 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010994 CAS Number: 10025-76-0 EINECS: 233-040-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...