Sodium bromate

Supplier: Apollo Scientific
Danger

IN1803-100G IN1803-25G
APOSIN1803-100GEA 29 EUR
APOSIN1803-100G APOSIN1803-25G
Sodium bromate
Sodium bromate
Formula: BrNaO₃
MW: 150,89 g/mol
Melting Pt: 381 °C (dec.)
Density: 3,33 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003476
CAS Number: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR