Holmium(III) chloride, anhydrous 99,9%

Supplier: Apollo Scientific
Warning

Synonyms: Holmium trichloride

IN1981-25G IN1981-5G
APOSIN1981-25GEA 522 EUR
APOSIN1981-25G APOSIN1981-5G
Holmium(III) chloride, anhydrous 99,9%
Holmium(III) chloride
Formula: Cl₃Ho
MW: 271,29 g/mol
Boiling Pt: 1500 °C
Melting Pt: 718 °C
Density: 3,7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011051
CAS Number: 10138-62-2
EINECS: 233-387-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR