Rhenium 99.999%, powder –22 mesh

Supplier: Apollo Scientific

IN2992-1G IN2992-5G
APOSIN2992-1GEA 144 EUR
APOSIN2992-1G APOSIN2992-5G
Rhenium 99.999%, powder –22 mesh
Rhenium
Formula: Re
MW: 186.21 g/mol
Boiling Pt: 5900 °C
Melting Pt: 3180 °C
Density: 21.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011195
CAS Number: 7440-15-5
Merck Index: 12,08343

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR