Ruthenium 99,95%, powder –200 mesh
Supplier: Apollo Scientific
|
Warning
|
IN3055-10G
IN3055-2G
APOSIN3055-10GEA
1280
EUR
APOSIN3055-10G
APOSIN3055-2G
Ruthenium 99,95%, powder –200 mesh
Ruthenium
Formula:
Ru MW: 101.07 g/mol Boiling Pt: 4900 °C Melting Pt: 2500 °C Density: 12.45 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 UN: 3089 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...