Terbium 99,9%, chips
Supplier: Apollo Scientific
|
Danger
|
IN3454-1G
IN3454-5G
APOSIN3454-1GEA
118
EUR
APOSIN3454-1G
APOSIN3454-5G
Terbium 99,9%, chips
Terbium
Formula:
Tb MW: 158,93 g/mol Boiling Pt: 2800 °C Melting Pt: 1356 °C Density: 8,234 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011256 CAS Number: 7440-27-9 UN: 3178 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...