Ytterbium(III) chloride hydrate 99.9+%
Supplier: Apollo Scientific
|
Warning
|
IN3799-10G
IN3799-50G
APOSIN3799-10GEA
58.2
EUR
APOSIN3799-10G
APOSIN3799-50G
Ytterbium(III) chloride hydrate 99.9+%
Ytterbium (III) chloride hydrate
Formula:
Cl₃Yb·H₂O MW: 297,41 g/mol Melting Pt: 150…155 °C Density: 2,575 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149884 CAS Number: 19423-87-1 EINECS: 233-800-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...