Zinc 99+%, mossy

Supplier: Apollo Scientific
Warning

IN3874-1KG IN3874-5KG
APOSIN3874-1KGEA 148 EUR
APOSIN3874-1KG APOSIN3874-5KG
Zinc 99+%, mossy
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908 °C
Melting Pt: 419 °C
Density: 7.133 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9+EHS III
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR