Zinc sulphide 99.99%
Supplier: Apollo Scientific
IN3893-250G
IN3893-50G
APOSIN3893-250GEA
626
EUR
APOSIN3893-250G
APOSIN3893-50G
Zinc sulphide 99.99%
Zinc sulphide
Formula:
SZn Melting Pt: 1700 °C Density: 4,1 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...